Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053600.2 | NP_446052.2 | 4 - 5 | 640 | 147 |
XM_006239638.2 | XP_006239700.1 | 4 - 5 | 691 | 147 | |
XM_006239639.2 | XP_006239701.1 | 4 - 5 | 691 | 147 | |
GenBank mRNA | AB076183.1 | - | 4 - 5 | 640 | 147 |
AB080086.1 | - | 4 - 5 | 619 | 147 | |
BC100060.1 | - | 4 - 5 | 621 | 147 | |
CK359698.1 | - | 4 - 5 | 607 | 147 | |
U64689.1 | - | 4 - 5 | 466 | 147 |