Assay Design
Gene Transcripts
- Gene Symbol
- Dnajc27
- Entrez Gene ID
-
298859
- Gene Name
- DnaJ (Hsp40) homolog, subfamily C, member 27
- Gene Aliases
-
Rbj
- Chromosome Location
-
Chr.6: 28515076 - 28544758 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY539931.1 |
- |
1 - 2 |
181 |
73 |
View Full Product Details