Assay Design
Gene Transcripts
- Gene Symbol
- Slc5a6
- Entrez Gene ID
-
170551
- Gene Name
- solute carrier family 5 (sodium/multivitamin and iodide cotransporter), member 6
- Gene Aliases
-
SMVT
- Chromosome Location
-
Chr.6: 26685823 - 26697110 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_130746.1 |
NP_570102.1 |
2 - 3 |
235 |
77 |
GenBank mRNA |
AF026554.1 |
- |
2 - 3 |
235 |
77 |
View Full Product Details