Assay Design



Gene Transcripts

Gene Symbol
Rtn1
Entrez Gene ID
116644
Gene Name
reticulon 1
Gene Aliases
Nsp
Chromosome Location
Chr.6: 94951016 - 95177477 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008764690.1 XP_008762912.1 1 - 2 205 60
GenBank mRNA FQ212949.1 - 1 - 2 116 60
FQ214343.1 - 1 - 2 41 60
U17603.1 - 1 - 2 175 60

View Full Product Details