Assay Design



Gene Transcripts

Gene Symbol
Rtn1
Entrez Gene ID
116644
Gene Name
reticulon 1
Gene Aliases
Nsp
Chromosome Location
Chr.6: 94951016 - 95177477 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053865.1 NP_446317.1 3 - 4 2182 121
GenBank mRNA U17604.1 - 3 - 4 2182 121

View Full Product Details