Assay Design



Gene Transcripts

Gene Symbol
Galm
Entrez Gene ID
313843
Gene Name
galactose mutarotase (aldose 1-epimerase)
Gene Aliases
-
Chromosome Location
Chr.6: 2808988 - 2860742 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007704.1 NP_001007705.1 3 - 4 607 92
GenBank mRNA BC081876.1 - 3 - 4 607 92

View Full Product Details