Assay Design



Gene Transcripts

Gene Symbol
Emilin1
Entrez Gene ID
298845
Gene Name
elastin microfibril interfacer 1
Gene Aliases
-
Chromosome Location
Chr.6: 26821249 - 26829010 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106710.1 NP_001100180.1 1 - 2 615 123
XM_006239793.2 XP_006239855.1 1 - 2 653 123

View Full Product Details