Assay Design



Gene Transcripts

Gene Symbol
Slc16a13
Entrez Gene ID
287451
Gene Name
solute carrier family 16, member 13
Gene Aliases
-
Chromosome Location
Chr.10: 56827593 - 56831243 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005530.1 NP_001005530.1 3 - 4 1362 105
GenBank mRNA BC081902.1 - 3 - 4 1362 105

View Full Product Details