Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031851.2 | NP_114039.1 | 1 - 2 | 35 | 63 |
GenBank mRNA | BC072518.1 | - | 1 - 2 | 35 | 63 |
FQ213801.1 | - | 1 - 2 | 50 | 63 | |
FQ229103.1 | - | 1 - 2 | 37 | 63 | |
FQ229278.1 | - | 1 - 2 | 70 | 63 | |
FQ233703.1 | - | 1 - 2 | 43 | 63 |