Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014122.1 | NP_001014144.1 | 2 - 3 | 212 | 82 |
XM_006247722.1 | XP_006247784.1 | 2 - 3 | 306 | 82 | |
XM_006247723.2 | XP_006247785.1 | 1 - 2 | 104 | 82 | |
GenBank mRNA | BC079310.1 | - | 2 - 3 | 212 | 82 |
FQ227422.1 | - | 2 - 3 | 306 | 82 |