Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007011.1 | NP_001007012.1 | 7 - 8 | 992 | 93 |
XM_006247384.2 | XP_006247446.1 | 7 - 8 | 2122 | 93 | |
XM_006247385.2 | XP_006247447.1 | 7 - 8 | 2049 | 93 | |
XM_006247387.2 | XP_006247449.1 | 6 - 7 | 1996 | 93 | |
GenBank mRNA | AY251605.1 | - | 7 - 8 | 984 | 93 |
AY251606.1 | - | 6 - 7 | 918 | 93 | |
BC082007.1 | - | 7 - 8 | 992 | 93 |