Assay Design



Gene Transcripts

Gene Symbol
Ttll6
Entrez Gene ID
287646
Gene Name
tubulin tyrosine ligase-like family, member 6
Gene Aliases
RGD1311922, Ttll6-ps1
Chromosome Location
Chr.10: 83965520 - 83999541 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008768194.1 XP_008766416.1 2 - 3 241 91
XM_008768195.1 XP_008766417.1 2 - 3 241 91

View Full Product Details