Assay Design



Gene Transcripts

Gene Symbol
Mmp25
Entrez Gene ID
302963
Gene Name
matrix metallopeptidase 25
Gene Aliases
-
Chromosome Location
Chr.10: 12962214 - 12966392 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008767528.1 XP_008765750.1 2 - 3 800 64

View Full Product Details