Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011982.1 | NP_001011982.1 | 6 - 7 | 1161 | 75 |
XM_006239888.2 | XP_006239950.1 | 7 - 8 | 1363 | 75 | |
XM_006239889.2 | XP_006239951.1 | 7 - 8 | 1360 | 75 | |
XM_006239890.2 | XP_006239952.1 | 7 - 8 | 1240 | 75 | |
XM_006239891.2 | XP_006239953.1 | 6 - 7 | 1186 | 75 | |
XM_006239892.2 | XP_006239954.1 | 6 - 7 | 1197 | 75 | |
GenBank mRNA | BC079437.1 | - | 6 - 7 | 1161 | 75 |