Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011982.1 | NP_001011982.1 | 4 - 5 | 872 | 66 |
XM_006239888.2 | XP_006239950.1 | 5 - 6 | 1074 | 66 | |
XM_006239889.2 | XP_006239951.1 | 5 - 6 | 1071 | 66 | |
XM_006239890.2 | XP_006239952.1 | 5 - 6 | 951 | 66 | |
XM_006239891.2 | XP_006239953.1 | 4 - 5 | 897 | 66 | |
XM_006239892.2 | XP_006239954.1 | 4 - 5 | 908 | 66 | |
GenBank mRNA | BC079437.1 | - | 4 - 5 | 872 | 66 |