Assay Design



Gene Transcripts

Gene Symbol
Aldoart2
Entrez Gene ID
299052
Gene Name
aldolase 1 A retrogene 2
Gene Aliases
Aldoal1, Aldoart1
Chromosome Location
Chr.6: 76349362 - 76351052 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013943.1 NP_001013965.1 1 - 1 1569 118
GenBank mRNA BC079243.1 - 1 - 1 1569 118

View Full Product Details