Assay Design



Gene Transcripts

Gene Symbol
Mbip
Entrez Gene ID
362740
Gene Name
MAP3K12 binding inhibitory protein 1
Gene Aliases
-
Chromosome Location
Chr.6: 77226439 - 77243695 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108712.2 NP_001102182.2 3 - 4 542 161
XM_006240120.2 XP_006240182.1 3 - 4 560 161
GenBank mRNA BC166982.1 - 3 - 4 469 161

View Full Product Details