Assay Design



Gene Transcripts

Gene Symbol
Mboat2
Entrez Gene ID
313997
Gene Name
membrane bound O-acyltransferase domain containing 2
Gene Aliases
Oact2
Chromosome Location
Chr.6: 44009872 - 44135501 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108016.2 NP_001101486.2 11 - 12 1295 108
XM_006239985.2 XP_006240047.1 10 - 11 1169 108
GenBank mRNA DV718174.1 - 1 - 2 117 108

View Full Product Details