Assay Design



Gene Transcripts

Gene Symbol
Pax9
Entrez Gene ID
362741
Gene Name
paired box 9
Gene Aliases
-
Chromosome Location
Chr.6: 77607705 - 77624453 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039539.1 NP_001034628.1 2 - 3 804 110
GenBank mRNA AB248957.1 - 2 - 3 804 110

View Full Product Details