Assay Design



Gene Transcripts

Gene Symbol
Tspan13
Entrez Gene ID
366602
Gene Name
tetraspanin 13
Gene Aliases
Tm4sf13
Chromosome Location
Chr.6: 55497659 - 55524415 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013244.1 NP_001013262.1 4 - 5 583 69
GenBank mRNA BC089906.1 - 4 - 5 583 69
FQ217050.1 - 4 - 5 598 69

View Full Product Details