Assay Design



Gene Transcripts

Gene Symbol
Nlrc4
Entrez Gene ID
298784
Gene Name
NLR family, CARD domain containing 4
Gene Aliases
Card12
Chromosome Location
Chr.6: 22167874 - 22194320 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008764491.1 XP_008762713.1 3 - 4 493 116
XM_008764492.1 XP_008762714.1 3 - 4 493 116
XM_008764493.1 XP_008762715.1 3 - 4 493 116

View Full Product Details