Assay Design



Gene Transcripts

Gene Symbol
Pdia6
Entrez Gene ID
286906
Gene Name
protein disulfide isomerase family A, member 6
Gene Aliases
CaBP1, P5, Txndc7
Chromosome Location
Chr.6: 42568269 - 42585654 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004442.1 NP_001004442.1 1 - 2 52 73
GenBank mRNA BC082063.1 - 1 - 2 52 73

View Full Product Details