Assay Design



Gene Transcripts

Gene Symbol
Sdr16c5
Entrez Gene ID
297805
Gene Name
short chain dehydrogenase/reductase family 16C, member 5
Gene Aliases
RGD1565999, Rdhe2
Chromosome Location
Chr.5: 16904926 - 16930076 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106634.1 NP_001100104.1 5 - 6 1059 121
XM_006237836.2 XP_006237898.1 6 - 7 946 121

View Full Product Details