Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007757.1 | NP_001007758.1 | 2 - 3 | 207 | 101 |
XM_006257228.2 | XP_006257290.1 | 3 - 4 | 328 | 101 | |
XM_008773423.1 | XP_008771645.1 | 6 - 7 | 1623 | 101 | |
XM_008773424.1 | XP_008771646.1 | 6 - 7 | 2625 | 101 | |
XM_008773425.1 | XP_008771647.1 | 3 - 4 | 352 | 101 | |
GenBank mRNA | BC081893.1 | - | 2 - 3 | 207 | 101 |