Assay Design



Gene Transcripts

Gene Symbol
Hnrnph2
Entrez Gene ID
308650
Gene Name
heterogeneous nuclear ribonucleoprotein H2 (H')
Gene Aliases
Hnrph2
Chromosome Location
Chr.X: 105417605 - 105423531 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014019.1 NP_001014041.1 3 - 4 171 64
XM_006257219.2 XP_006257281.1 3 - 4 214 64
GenBank mRNA BC079240.1 - 3 - 4 171 64

View Full Product Details