Assay Design



Gene Transcripts

Gene Symbol
Muc13
Entrez Gene ID
207126
Gene Name
mucin 13, cell surface associated
Gene Aliases
Ly64, MUC-13, Muc-123
Chromosome Location
Chr.11: 70185757 - 70223012 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008768788.1 XP_008767010.1 11 - 12 1691 67
GenBank mRNA U89744.1 - 10 - 11 1653 67

View Full Product Details