Assay Design



Gene Transcripts

Gene Symbol
Wdr53
Entrez Gene ID
498097
Gene Name
WD repeat domain 53
Gene Aliases
RGD1559546
Chromosome Location
Chr.11: 71787454 - 71796258 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109055.1 NP_001102525.1 1 - 2 347 108

View Full Product Details