Assay Design



Gene Transcripts

Gene Symbol
Golgb1
Entrez Gene ID
192243
Gene Name
golgin B1
Gene Aliases
-
Chromosome Location
Chr.11: 66761645 - 66820989 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138885.1 NP_620240.1 16 - 17 9096 106
XM_006248379.2 XP_006248441.1 17 - 18 11089 106
XM_006248380.2 XP_006248442.1 16 - 17 10937 106
XM_008768698.1 XP_008766920.1 16 - 17 10282 106
XM_008768699.1 XP_008766921.1 16 - 17 10267 106
XM_008768700.1 XP_008766922.1 16 - 17 10264 106
XM_008768701.1 XP_008766923.1 15 - 16 10159 106
XM_008768702.1 XP_008766924.1 14 - 15 8808 106
GenBank mRNA D25543.1 - 16 - 17 9096 106

View Full Product Details