Assay Design



Gene Transcripts

Gene Symbol
Golgb1
Entrez Gene ID
192243
Gene Name
golgin B1
Gene Aliases
-
Chromosome Location
Chr.11: 66761645 - 66820989 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138885.1 NP_620240.1 18 - 19 9376 82
XM_006248379.2 XP_006248441.1 19 - 20 11369 82
XM_006248380.2 XP_006248442.1 18 - 19 11217 82
XM_008768698.1 XP_008766920.1 18 - 19 10562 82
XM_008768699.1 XP_008766921.1 18 - 19 10547 82
XM_008768700.1 XP_008766922.1 18 - 19 10544 82
XM_008768701.1 XP_008766923.1 17 - 18 10439 82
XM_008768702.1 XP_008766924.1 16 - 17 9088 82
GenBank mRNA D25543.1 - 18 - 19 9376 82

View Full Product Details