Assay Design



Gene Transcripts

Gene Symbol
Gpc3
Entrez Gene ID
25236
Gene Name
glypican 3
Gene Aliases
OCI-5
Chromosome Location
Chr.X: 139579268 - 139947093 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012774.1 NP_036906.1 5 - 6 1403 98
GenBank mRNA BC085756.1 - 5 - 6 1418 98
M22400.1 - 5 - 6 1403 98

View Full Product Details