Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012518.3 | NP_036650.1 | 4 - 5 | 432 | 83 |
GenBank mRNA | BC063187.1 | - | - | 527 | 83 |
FQ212716.1 | - | - | 498 | 83 | |
M16659.1 | - | 4 - 5 | 432 | 83 | |
X13817.1 | - | 5 - 6 | 524 | 83 |