Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106226.2 | NP_001099696.2 | 7 - 8 | 3100 | 65 |
XM_006228343.2 | XP_006228405.1 | 9 - 10 | 3447 | 65 | |
XM_006228344.2 | XP_006228406.1 | 8 - 9 | 3381 | 65 | |
XM_006228345.2 | XP_006228407.1 | 8 - 9 | 3270 | 65 |