Assay Design



Gene Transcripts

Gene Symbol
Vrk3
Entrez Gene ID
361565
Gene Name
vaccinia related kinase 3
Gene Aliases
-
Chromosome Location
Chr.1: 100755621 - 100783008 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005561.1 NP_001005561.1 13 - 14 1304 60
XM_006229095.2 XP_006229157.1 13 - 14 1363 60
GenBank mRNA BC081928.1 - 13 - 14 1304 60

View Full Product Details