Assay Design



Gene Transcripts

Gene Symbol
Klk10
Entrez Gene ID
292850
Gene Name
kallikrein related-peptidase 10
Gene Aliases
-
Chromosome Location
Chr.1: 99718433 - 99722595 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004100.1 NP_001004100.1 4 - 5 683 99
XM_006228987.2 XP_006229049.1 5 - 6 818 99

View Full Product Details