Assay Design



Gene Transcripts

Gene Symbol
Sult2b1
Entrez Gene ID
292915
Gene Name
sulfotransferase family, cytosolic, 2B, member 1
Gene Aliases
ST2B1
Chromosome Location
Chr.1: 101712254 - 101773392 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006229031.2 XP_006229093.1 1 - 2 149 115
GenBank mRNA AY827148.1 - 1 - 2 117 115

View Full Product Details