Assay Design
Gene Transcripts
- Gene Symbol
- Iglon5
- Entrez Gene ID
-
308557
- Gene Name
- IgLON family member 5
- Gene Aliases
-
RGD1305344
- Chromosome Location
-
Chr.1: 98440186 - 98457942 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_218634.8 |
XP_218634.5 |
6 - 7 |
766 |
78 |
View Full Product Details