Assay Design



Gene Transcripts

Gene Symbol
Lrfn1
Entrez Gene ID
365222
Gene Name
leucine rich repeat and fibronectin type III domain containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 85290887 - 85308518 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108910.1 NP_001102380.1 2 - 3 92 154
XM_006228626.2 XP_006228688.1 3 - 4 1459 154
XM_006228628.2 XP_006228690.1 3 - 4 762 154
XM_008759173.1 XP_008757395.1 2 - 3 1760 154
XM_008759174.1 XP_008757396.1 3 - 4 1851 154
XM_008759175.1 XP_008757397.1 3 - 4 583 154

View Full Product Details