Assay Design
Gene Transcripts
- Gene Symbol
- Atp1a3
- Entrez Gene ID
-
24213
- Gene Name
- ATPase, Na+/K+ transporting, alpha 3 polypeptide
- Gene Aliases
-
Atpa1a3
- Chromosome Location
-
Chr.1: 81852425 - 81881549 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_012506.1 |
NP_036638.1 |
1 - 2 |
147 |
66 |
GenBank mRNA |
M14513.1 |
- |
1 - 2 |
147 |
66 |
View Full Product Details