Assay Design



Gene Transcripts

Gene Symbol
Rabac1
Entrez Gene ID
83583
Gene Name
Rab acceptor 1 (prenylated)
Gene Aliases
Pra1
Chromosome Location
Chr.1: 81843667 - 81846765 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031774.1 NP_113962.1 4 - 5 566 60
GenBank mRNA AF025506.1 - 4 - 5 566 60
BC086387.1 - 4 - 5 533 60
FQ210487.1 - 4 - 5 559 60

View Full Product Details