Assay Design



Gene Transcripts

Gene Symbol
Gbas
Entrez Gene ID
498174
Gene Name
glioblastoma amplified sequence
Gene Aliases
-
Chromosome Location
Chr.12: 30527540 - 30566080 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017486.1 NP_001017486.1 4 - 5 360 57
XM_006249280.2 XP_006249342.1 4 - 5 421 57
XM_008769201.1 XP_008767423.1 4 - 5 422 57
GenBank mRNA BC086385.1 - 4 - 5 360 57

View Full Product Details