Assay Design



Gene Transcripts

Gene Symbol
Idh2
Entrez Gene ID
361596
Gene Name
isocitrate dehydrogenase 2 (NADP+), mitochondrial
Gene Aliases
-
Chromosome Location
Chr.1: 141874354 - 141893674 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014161.1 NP_001014183.1 1 - 2 147 81
GenBank mRNA BC076398.1 - 1 - 2 147 81

View Full Product Details