Assay Design



Gene Transcripts

Gene Symbol
Lysmd4
Entrez Gene ID
681647
Gene Name
LysM, putative peptidoglycan-binding, domain containing 4
Gene Aliases
-
Chromosome Location
Chr.1: 128199113 - 128204989 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008759576.1 XP_008757798.1 3 - 4 501 109

View Full Product Details