Assay Design



Gene Transcripts

Gene Symbol
Eftud1
Entrez Gene ID
308789
Gene Name
elongation factor Tu GTP binding domain containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 144601425 - 144740634 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107534.1 NP_001101004.1 9 - 10 1000 90
GenBank mRNA FQ221869.1 - 2 - 3 162 90

View Full Product Details