Assay Design



Gene Transcripts

Gene Symbol
Fbxo25
Entrez Gene ID
364637
Gene Name
F-box protein 25
Gene Aliases
-
Chromosome Location
Chr.16: 80770788 - 80806661 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014239.1 NP_001014261.1 3 - 4 271 130
GenBank mRNA BC082088.1 - 3 - 4 271 130

View Full Product Details