Assay Design



Gene Transcripts

Gene Symbol
Adrb3
Entrez Gene ID
25645
Gene Name
adrenoceptor beta 3
Gene Aliases
ADRB
Chromosome Location
Chr.16: 69003543 - 69006632 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013108.2 NP_037240.2 2 - 3 1453 131
GenBank mRNA M74716.1 - 2 - 3 1452 131
S56481.1 - 2 - 3 1244 131
S73473.1 - 2 - 3 1283 131

View Full Product Details