Assay Design



Gene Transcripts

Gene Symbol
Peli3
Entrez Gene ID
309157
Gene Name
pellino E3 ubiquitin protein ligase family member 3
Gene Aliases
RGD1305989
Chromosome Location
Chr.1: 220193403 - 220204032 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127542.1 NP_001121014.1 2 - 3 290 75
GenBank mRNA BC161922.1 - 2 - 3 276 75

View Full Product Details