Assay Design



Gene Transcripts

Gene Symbol
Dnajc4
Entrez Gene ID
361717
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 4
Gene Aliases
-
Chromosome Location
Chr.1: 222242889 - 222247110 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013196.1 NP_001013214.1 3 - 4 319 97
XM_006230859.2 XP_006230921.1 3 - 4 314 97
XM_006230860.2 XP_006230922.1 3 - 4 276 97
XM_006230861.2 XP_006230923.1 2 - 3 474 97
XM_006230862.2 XP_006230924.1 2 - 3 494 97
XM_008760159.1 XP_008758381.1 3 - 4 313 97
GenBank mRNA BC088201.1 - 3 - 4 319 97

View Full Product Details