Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013196.1 | NP_001013214.1 | 5 - 6 | 670 | 62 |
XM_006230859.2 | XP_006230921.1 | 5 - 6 | 662 | 62 | |
XM_006230860.2 | XP_006230922.1 | 5 - 6 | 627 | 62 | |
XM_006230861.2 | XP_006230923.1 | 4 - 5 | 825 | 62 | |
XM_006230862.2 | XP_006230924.1 | 4 - 5 | 845 | 62 | |
XM_008760159.1 | XP_008758381.1 | 5 - 6 | 664 | 62 | |
GenBank mRNA | BC088201.1 | - | 5 - 6 | 670 | 62 |