Assay Design



Gene Transcripts

Gene Symbol
Dnajc4
Entrez Gene ID
361717
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 4
Gene Aliases
-
Chromosome Location
Chr.1: 222242889 - 222247110 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013196.1 NP_001013214.1 6 - 7 756 73
XM_006230859.2 XP_006230921.1 6 - 7 748 73
XM_006230860.2 XP_006230922.1 6 - 7 713 73
XM_006230861.2 XP_006230923.1 5 - 6 911 73
XM_006230862.2 XP_006230924.1 5 - 6 931 73
XM_008760159.1 XP_008758381.1 6 - 7 750 73
GenBank mRNA BC088201.1 - 6 - 7 756 73

View Full Product Details