Assay Design



Gene Transcripts

Gene Symbol
Ms4a1
Entrez Gene ID
309217
Gene Name
membrane-spanning 4-domains, subfamily A, member 1
Gene Aliases
-
Chromosome Location
Chr.1: 227429241 - 227441493 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107578.1 NP_001101048.1 4 - 5 591 154
XM_006231089.2 XP_006231151.1 5 - 6 763 154
GenBank mRNA FQ225563.1 - 5 - 6 154
FQ235337.1 - 4 - 5 528 154

View Full Product Details